Anti-MDM2 (Ab-2) (2A10) Mouse Monoclonal Antibody [clone: 2A10]
Catalog # OP115-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:MDM2
- Clonality:Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MDM2 (Ab-2) (2A10)
Clonality: monoclonal
Clone: 2A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human