Order Entry
Italy
ContactUsLinkComponent
Anti-MMP-2 (Ab-3) (42-5D11) Mouse Monoclonal Antibody
Anti-MMP-2 (Ab-3) (42-5D11) Mouse Monoclonal Antibody
Catalog # IM33-100
Anti-MMP-2 (Ab-3) (42-5D11) Mouse Monoclonal Antibody
Catalog # IM33-100

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MMP-2
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: MMP-2 (Ab-3) (42-5D11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: