Anti-Cathepsin B (Ab-1) (CA10) Mouse Monoclonal Antibody [clone: CA10]
IM27L-100
- Antibody type:Primary
- Antigen name:Cathepsin B
- Antigen symbol:CTSB
- Clonality:Monoclonal
- Clone:CA10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Cathepsin B (Ab-1) (CA10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: