Order Entry
Italy
ContactUsLinkComponent
Anti-VASP Mouse Monoclonal Antibody [clone: 5C6]
Anti-VASP Mouse Monoclonal Antibody [clone: 5C6]
Catalog # 676604-100
Anti-VASP Mouse Monoclonal Antibody [clone: 5C6]
Catalog # 676604-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vasodilator-stimulated phosphoprotein
  • Antigen symbol:
    VASP
  • Clonality:
    Monoclonal
  • Clone:
    5C6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: VASP (vasodilator-stimulated phosphoprotein)
Clonality: monoclonal
Clone: 5C6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: