Anti-MAPT Rabbit Polyclonal Antibody
Catalog # 577815-10
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:microtubule-associated Protein tau
- Antigen symbol:MAPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:10 Tests
- Pk:10 Tests
Specifications
About this item
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human