Anti-p38 MAP Kinase Rabbit Polyclonal Antibody
506119-10
- Antibody type:Primary
- Antigen name:p38 MAP Kinase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:10 Tests
- Pk:10 Tests
Type: Primary
Antigen: p38 MAP Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: