Order Entry
Italy
ContactUsLinkComponent
Anti-NMDAR 2B (984-1104) Rabbit Polyclonal Antibody
Anti-NMDAR 2B (984-1104) Rabbit Polyclonal Antibody
Catalog # 454582-10
Anti-NMDAR 2B (984-1104) Rabbit Polyclonal Antibody
Catalog # 454582-10

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    NMDANR2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    10 µg
  • Pk:
    10 µG

Specifications

About this item

Type: Primary
Antigen: NMDAR 2B (984-1104)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: