Order Entry
Italy
ContactUsLinkComponent
Anti-beta-Interferon Rabbit Polyclonal Antibody
Anti-beta-Interferon Rabbit Polyclonal Antibody
Catalog # 407299-20
Anti-beta-Interferon Rabbit Polyclonal Antibody
Catalog # 407299-20

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta-Interferon
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    20000 Active Unit
  • Pk:
    20 KU

Specifications

About this item

Type: Primary
Antigen: beta-Interferon
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human