Anti-alpha-Interferon (MMHA-2) Mouse Monoclonal Antibody
407290-500
- Antibody type:Primary
- Antigen name:alpha Interferon
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: alpha-Interferon (MMHA-2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human