Anti-alpha-Epithelial Cell Sodium Channel (20-42) Rabbit Polyclonal Antibody
324870-100
- Antibody type:Primary
- Antigen name:alpha Epithelial Cell Sodium Channel
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: alpha-Epithelial Cell Sodium Channel (20-42)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: