Anti-Dopamine D 5 Receptor Rabbit Polyclonal Antibody
Catalog # 324408-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:Dopamine D5 receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Dopamine D 5 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: