Anti-GFP Rabbit polyclonal antibody unconjugated
GENSA01704-100
- Antibody type:Tag
- Antigen name:Green Fluorescence Protein
- Antigen symbol:GFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Antigen synonyms:GFPuv|eGFP|Green Fluorescent Protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: