Anti-HA tag Mouse Monoclonal Antibody (FITC (Fluorescein))
Catalog # GENSA01621
Supplier: Genscript
Specifications
- Antibody type:Tag
- Antigen name:HA tag
- Antigen symbol:HA
- Clonality:Monoclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: