Order Entry
Italy
ContactUsLinkComponent
Anti-beta-Amyloid Rabbit polyclonal antibody
Anti-beta-Amyloid Rabbit polyclonal antibody
Catalog # GENSA01567-40
Supplier:  Genscript
undefined
Anti-beta-Amyloid Rabbit polyclonal antibody
Catalog # GENSA01567-40
Supplier:  Genscript

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta Amyloid
  • Antigen symbol:
    A beta
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Antigen synonyms:
    APP|CTFgamma|CVAP|ABETA|APPI|AD1|Peptidase nexin-II|amyloid beta (A4) precursor protein|Alzheimer disease amyloid A4 protein homolog|ABPP|Amyloid beta A4 protein|Amyloidogenic glycoprotein AG|AAA|PN2
  • Amino acid number:
    1 to 42
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    40 µg
  • Pk:
    40 µG

Specifications

About this item

Type: Primary
Antigen: ?-amyloid
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: