Order Entry
Italy
ContactUsLinkComponent
Rabbit IgG Isotype Control (Unconjugated)
Rabbit IgG Isotype Control (Unconjugated)
Catalog # GENSA01008
Supplier:  Genscript
Rabbit IgG Isotype Control (Unconjugated)
Catalog # GENSA01008
Supplier:  Genscript

Specifications

  • Antibody type:
    Control
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    4 mg
  • Pk:
    4 mg

Specifications

About this item

Type: Control
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: