Anti-E-tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # GENSA00874-40
Supplier: Genscript
Specifications
- Antibody type:Tag
- Antigen symbol:E-tag
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:40 µg
- Pk:40 µG
Specifications
About this item
Type: Tag
Antigen: E-tag
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: