Anti-GST Goat Polyclonal Antibody
Catalog # 27-4577-01
Supplier: Cytiva
Specifications
- Antibody type:Tag
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Multiple
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µl
- Pk:0,5 mL
Specifications
About this item
Anti-GST Antibody is a polyclonal antibody purified from the sera of goats immunized with purified schistosomal GST, Because of its polyclonal nature, it can recognize more than one epitope on GST.
- For the highly sensitive and specific detection of recombinant schistosomal GST fusion proteins expressed using pGEX vectors
- Extensively cross-adsorbed to remove antibodies that bind to E. coli proteins
- Supplied unconjugated for use with any enzyme-conjugated anti-goat antibody
- Recommended for use in Western blots and dot blots to detect GST fusion proteins
Because of its polyclonal nature, it can recognize more than one epitope on GST, thereby improving its capacity for recognizing GST fusion proteins even if some binding sites are masked due to recombinant protein folding.
Type: Tag
Antigen: GST (Glutathione-S-Transferase)
Clonality: polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Pan