Anti-Npc1l1 (aa56-69) Goat Polyclonal Antibody
Catalog # EVBIEB10837
Nome del fornitore: Everest Biotech
Specifiche
- Antibody type:Primary
- Antigen name:Niemann-Pick C1-Like 1
- Antigen symbol:NPC1L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:N-terminal
- Cross adsorption:none
- Amino acid number:56 to 69
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: Npc1l1 (aa56-69)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat