Anti-SLC12A2 Goat Polyclonal Antibody
Catalog # EVBIEB09783
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 12 (sodium/potassium/chloride transporter), member 2
- Antigen symbol:Solute carrier family 12, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SLC12A2 (solute carrier family 12 (sodium/potassium/chloride transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: