Anti-KLRG1 Goat Polyclonal Antibody
Catalog # EVBIEB09346
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:killer cell lectin-like receptor subfamily G, member 1
- Antigen symbol:KLRG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: KLRG1 (killer cell lectin-like receptor subfamily G, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human