Order Entry
Italy
ContactUsLinkComponent
Anti-STK36 Goat Polyclonal Antibody
Anti-STK36 Goat Polyclonal Antibody
Catalog # EVBIEB09334
Supplier:  Everest Biotech
Anti-STK36 Goat Polyclonal Antibody
Catalog # EVBIEB09334
Supplier:  Everest Biotech

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serine/threonine Kinase 36
  • Antigen symbol:
    STK36
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Dog
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: STK36 (serine/threonine kinase 36)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human