Anti-PGC-1beta / PERC Goat Polyclonal Antibody
Catalog # EVBIEB06764
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen symbol:PGC-1 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Epitope:N-terminal
- Cross adsorption:none
- Antigen synonyms:PERC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: PGC-1beta / PERC
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat