Anti-Collagen IV Rabbit Polyclonal Antibody
Catalog # BSBTPA1521
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Collagen 4
- Antigen symbol:COL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:COL4A1 NC1 domain,collagen IV|collagen alpha-1(IV) chain|collagen of basement membrane|alpha-1 chain|alpha-1 polypeptide
- Amino acid number:1697 to 1712
- Sequence:LIRTHISRCQVCMKNL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Collagen IV (1697-1712aa LIRTHISRCQVCMKNL), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Collagen alpha-2(IV) chain(COL4A2) detection. Tested with WB, IHC-P, IHC-F, ICC in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Collagen IV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat