Anti-p16INK4a/CDKN2 Mouse Monoclonal Antibody [clone: IMD-16]
BSBTMA1074
- Antibody type:Primary
- Antigen symbol:p16INK4a/CDKN2
- Clonality:Monoclonal
- Clone:IMD-16
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:p16INK4a
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant human p16 protein.
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: p16INK4a/CDKN2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human