Order Entry
Italy
ContactUsLinkComponent
 
Anti-Episialin, EMA Mouse Monoclonal Antibody [clone: EMA-39]
  BSBTMA1039
undefined
Anti-Episialin, EMA Mouse Monoclonal Antibody [clone: EMA-39]
  BSBTMA1039

 

  • Antibody type:
    Primary
  • Antigen name:
    Episialin, EMA
  • Antigen symbol:
    EMA
  • Clonality:
    Monoclonal
  • Clone:
    EMA-39
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Human milk fat globule membranes.
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Mouse IgG monoclonal antibody for Episialin, EMA, mucin 1, cell surface associated (MUC1) detection. Tested with IHC-P, IHC-F in Human. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Episialin, EMA
Clonality: Monoclonal
Clone: EMA-39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human