Anti-Leukotriene B4 Receptor 1 Rabbit Polyclonal Antibody
NOVUNBP2-27422
- Antibody type:Primary
- Antigen name:leukotriene B4 receptor
- Antigen symbol:LTB4R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:1241
- Antigen synonyms:LTBR1|LTB4R1|GPR16|CMKRL1|P2Y7|P2RY7|BLT1|BLTR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: Leukotriene B4 Receptor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: