Anti-AP3D1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27158
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 3, delta 1 subunit
- Antigen symbol:AP3D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:8943
- Antigen synonyms:ADTD|hBLVR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: AP3D1 (adaptor-related protein complex 3, delta 1 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human