Anti-NOD2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27118
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Nucleotide binding oligomerization domain containing 2
- Antigen symbol:NOD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:64127
- Antigen synonyms:NLRC2|PSORAS1|CARD15|CD|IBD1|ACUG|NOD2B|CLR16.3|BLAU
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: NOD2 (nucleotide-binding oligomerization domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human