Anti-HMGB1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27117
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:high mobility group box 1
- Antigen symbol:HMGB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:3146
- Antigen synonyms:SBP-1|HMG3|HMG1|HMG-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human