Anti-HES2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27110
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Hes family bHLH transcription factor 2
- Antigen symbol:HES2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Gene ID:54626
- Antigen synonyms:bHLHb40
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: HES2 (hes family bHLH transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: