Anti-HOXD8 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27108
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:homeobox D8
- Antigen symbol:HOXD8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:3234
- Antigen synonyms:HOX5.4|HOX4E|HOX4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: HOXD8 (homeobox D8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: