Order Entry
Italy
ContactUsLinkComponent
 
Anti-CLEC4E Rabbit Polyclonal Antibody
  NOVUNBP2-27093
undefined
Anti-CLEC4E Rabbit Polyclonal Antibody
  NOVUNBP2-27093

 

  • Antibody type:
    Primary
  • Antigen name:
    C-type lectin domain family 4, member E
  • Antigen symbol:
    CLEC4E
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Gene ID:
    26253
  • Antigen synonyms:
    MINCLE|CLECSF9
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: CLEC4E (C-type lectin domain family 4, member E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: