Anti-CLEC9A Mouse Monoclonal Antibody [clone: 14N8D7]
Catalog # NOVUNBP2-27087
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:C-type lectin domain family 9, member A
- Antigen symbol:CLEC9A
- Clonality:Monoclonal
- Clone:14N8D7
- Conjugation:Unconjugated
- Host:Mouse
- Western blot:Yes
- Gene ID:283420
- Antigen synonyms:DNGR1|UNQ9341|DNGR-1|CD370
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CLEC9A (C-type lectin domain family 9, member A)
Clonality: monoclonal
Clone: 14N8D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: