Anti-14-3-3 sigma/Stratifin Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27072
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:stratifin
- Antigen symbol:SFN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:2810
- Antigen synonyms:YWHAS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: 14-3-3 sigma/Stratifin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: