Anti-UNC93B1 Rabbit Polyclonal Antibody
NOVUNBP2-24743
- Antibody type:Primary
- Antigen name:unc-93 homolog B1 (C. elegans)
- Antigen symbol:UNC93B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:81622
- Antigen synonyms:Unc-93B1|UNC93B|IIAE1|UNC93
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: UNC93B1 (unc-93 homolog B1 (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat