Anti-ACAP2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24528
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
- Antigen symbol:ACAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:23527
- Antigen synonyms:CNT-B2|CENTB2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ACAP2 (ArfGAP with coiled-coil, ankyrin repeat and PH domains 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: