Order Entry
Italy
ContactUsLinkComponent
Anti-NR2F6 Rabbit Polyclonal Antibody
Anti-NR2F6 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24510
Supplier:  Novus Biologicals
undefined
Anti-NR2F6 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24510
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    nuclear receptor subfamily 2, group F, member 6
  • Antigen symbol:
    NR2F6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Gene ID:
    2063
  • Antigen synonyms:
    EAR2|EAR-2|ERBAL2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: NR2F6 (nuclear receptor subfamily 2, group F, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: