Anti-NOS2 Mouse Monoclonal Antibody [clone: 4E5]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: monoclonal
Clone: 400000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP2-22119SS
- NOVUNBP2-22119
- Antigen symbol:
- NOS2
- NOS2
- Antigen name:
- Nitric Oxide Synthase 2
- Nitric Oxide Synthase 2
- Antigen synonyms:
- INOS|HEP-NOS|NOS2A|NOS
- INOS|HEP-NOS|NOS2A|NOS
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 4E5
- 4E5
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 4843
- 4843
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together