Anti-LAMP2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: H4B4]
NOVUNBP2-21799
- Antibody type:Primary
- Antigen name:Lysosomal-associated membrane protein 2
- Antigen symbol:LAMP2
- Clonality:Monoclonal
- Clone:H4B4
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:3920
- Antigen synonyms:LAMP-2|LGP-B|Lysosome-associated membrane protein 2|CD107 antigen-like family member B|LAMP2|Lysosome-associated membrane glycoprotein 2|CD107b|Lysosomal membrane glycoprotein type B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: LAMP2 (lysosomal-associated membrane protein 2)
Clonality: monoclonal
Clone: H4B4
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human