Mouse IgG2a Isotype Control [clone: C1.18.4]
Catalog # NOVUNBP2-14864
Supplier: Novus Biologicals
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:C1.18.4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Control
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: monoclonal
Clone: C1.18.4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse