Anti-RBFOX1 Mouse Monoclonal Antibody (DyLight® 650) [clone: D8H8]
Catalog # NOVUNBP2-13169C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:RNA Binding Protein, Fox-1 homolog 1
- Antigen symbol:RBFOX1
- Clonality:Monoclonal
- Clone:D8H8
- Conjugation:DyLight® 650
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:54715
- Antigen synonyms:2BP1|FOX1|HRNBP1|FOX-1|A2BP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: RBFOX1 (RNA binding protein, fox-1 homolog (C. elegans) 1)
Clonality: monoclonal
Clone: D8H8
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse