Anti-PIF1 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-97931V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:PIF1 5'-to-3' DNA helicase
- Antigen symbol:PIF1
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Reactivity:Mouse
- Gene ID:80119
- Antigen synonyms:PIF|C15orf20
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: PIF1 (PIF1 5'-to-3' DNA helicase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse