Anti-TRPV1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNBP1-97417F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:transient receptor potential cation channel, subfamily V, member 1
- Antigen symbol:TRPV1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:7442
- Antigen synonyms:VR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TRPV1 (transient receptor potential cation channel, subfamily V, member 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: