Order Entry
Italy
ContactUsLinkComponent
Anti-SERF1A Rabbit Polyclonal Antibody
Anti-SERF1A Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-SERF1A Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-78392
  • NOVUNBP1-78392SS
  • Antigen symbol:
  • SERF1A
  • SERF1A
  • Antigen name:
  • Small EDRK-rich Factor 1A
  • Small EDRK-rich Factor 1A
  • Antigen synonyms:
  • H4F5|SERF1|FAM2A|4F5|SMAM1
  • H4F5|SERF1|FAM2A|4F5|SMAM1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 8293
  • 8293
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together