Anti-ABCB5 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-77689G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, Subfamily B, Member 5
- Antigen symbol:ABCB5
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Gene ID:340273
- Antigen synonyms:P-glycoprotein ABCB5|ATP-binding cassette sub-family B member 5|ABCB5 P-gp|ABCB5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ABCB5 (ATP-binding cassette, sub-family B (MDR/TAP), member 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human