Anti-IGF1R Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-77680R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Insulin-Like Growth Factor 1 Receptor
- Antigen symbol:IGF1R
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Gene ID:3480
- Antigen synonyms:MGC18216|JTK13|CD221|IGFIR|MGC142170|IGF1R|MGC142172
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: