Anti-TPH2 Rabbit Polyclonal Antibody (DyLight® 488)
NOVUNBP1-77556G
- Antibody type:Primary
- Antigen name:Tryptophan hydroxylase 2
- Antigen symbol:TPH2
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:121278
- Antigen synonyms:tryptophan 5-hydroxylase 2|neuronal tryptophan hydroxylase|tryptophan 5-monooxygenase 2|NTPH|ADHD7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: TPH2 (tryptophan hydroxylase 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human