Anti-TNFAIP3 Mouse Monoclonal Antibody [clone: 59A426]
NOVUNBP1-77533
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Alpha-induced Protein 3
- Antigen symbol:TNFAIP3
- Clonality:Monoclonal
- Clone:59A426
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:7128
- Antigen synonyms:A20p37|A20p50|OTU domain-containing protein 7C|Tumor necrosis factor alpha-induced protein 3|OTUD7C|TNF alpha-induced protein 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
The A20 / TNFAIP3 Antibody (59A426) from Novus Biologicals is a mouse monoclonal antibody to A20 / TNFAIP3. This antibody reacts with human, mouse, rat. The A20 / TNFAIP3 Antibody (59A426) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
Simple Western , CyTOF-ready
Type: Primary
Antigen: A20/TNFAIP3
Clonality: Monoclonal
Clone: 59A426
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat