Anti-HMOX1 Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNBP1-77459V
- Antibody type:Primary
- Antigen name:Heme Oxidase 1
- Antigen symbol:HMOX1
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:3162
- Antigen synonyms:HSP32|bK286B10|HO-1|HMOX1D
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: HMOX1 (heme oxygenase (decycling) 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse