Anti-COL1 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-77457R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Collagen, type I, alpha 1
- Antigen symbol:COL1A1
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human,Mouse
- Gene ID:1277
- Antigen synonyms:EDSC|OI4|OI2|OI3|OI1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse